2-Fluorobenzaldehyde 99%
Supplier: Apollo Scientific
Warning
PC3450-100G PC3450-1KG PC3450-250G
APOSPC3450-100GEA 24.8 EUR
APOSPC3450-100G APOSPC3450-1KG APOSPC3450-250G
2-Fluorobenzaldehyde 99%
2-Fluorobenzaldehyde
Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 172…175 °C
Melting Pt: –45 °C
Density: 1.178 g/cm³ (25 °C)
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00003302
CAS Number: 446-52-6
EINECS: 207-171-2
UN: 1989
ADR: 3,III