4-Fluoro-2-iodoaniline 98%
Supplier: Apollo Scientific
Warning
PC3452-100G PC3452-25G PC3452-5G
APOSPC3452-100GEA 102 EUR
APOSPC3452-100G APOSPC3452-25G APOSPC3452-5G
4-Fluoro-2-iodoaniline 98%
4-Fluoro-2-iodoaniline
Formula: C₆H₅FIN
MW: 237.02 g/mol
Boiling Pt: 74…76 °C
Density: 1.99 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042584
CAS Number: 61272-76-2
UN: 2810
ADR: 6.1 III