4'-Hydroxy-2'-(trifluoromethyl)acetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Hydroxy-2-(trifluoromethyl)phenyl)ethanone
PC3457-1G PC3457-25G PC3457-5G
APOSPC3457-1GEA 27.7 EUR
APOSPC3457-1G APOSPC3457-25G APOSPC3457-5G
4'-Hydroxy-2'-(trifluoromethyl)acetophenone
4'-Hydroxy-2'-(trifluoromethyl)acetophenone
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01091004
CAS Number: 220227-53-2