6-Fluoro-o-anisaldehyde 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-6-mehoxybenzaldehyde, 2-Fluoro-6-methoxybenzaldehyde, 6-Fluoro-2-anisaldehyde
PC3458-1G PC3458-25G PC3458-5G
APOSPC3458-1GEA 21.9 EUR
APOSPC3458-1G APOSPC3458-25G APOSPC3458-5G
6-Fluoro-o-anisaldehyde 98%
6-Fluoro-o-anisaldehyde
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 59…63 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01090998
CAS Number: 146137-74-8