3-Fluorobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC3460-100G PC3460-1KG PC3460-500G
APOSPC3460-100GEA 23.2 EUR
APOSPC3460-100G APOSPC3460-1KG APOSPC3460-500G
3-Fluorobenzaldehyde 98%
3-Fluorobenzaldehyde
Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 56…58 °C/10mmHg
Density: 1.174 g/cm³ (25 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00003348
CAS Number: 456-48-4
EINECS: 207-266-9
UN: 1989
ADR: 3,III