3-Fluorobenzamide 99%
Supplier: Apollo Scientific
Warning
PC3474-100G PC3474-25G
APOSPC3474-100GEA 159 EUR
APOSPC3474-100G APOSPC3474-25G
3-Fluorobenzamide 99%
3-Fluorobenzamide
Formula: C₇H₆FNO
MW: 139.13 g/mol
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00007983
CAS Number: 455-37-8
EINECS: 207-247-5