4-Methoxy-2-(trifluoromethyl)benzaldehyde
Supplier: Apollo Scientific
Warning
PC3479-1G PC3479-25G PC3479-5G
APOSPC3479-1GEA 21.9 EUR
APOSPC3479-1G APOSPC3479-25G APOSPC3479-5G
4-Methoxy-2-(trifluoromethyl)benzaldehyde
4-Methoxy-2-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Melting Pt: 39…41 °C
Storage Temperature: Ambient
MDL Number: MFCD01091011
CAS Number: 106312-36-1