3-Fluorobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC3510-100G PC3510-1KG PC3510-500G
APOSPC3510-100GEA 21.9 EUR
APOSPC3510-100G APOSPC3510-1KG APOSPC3510-500G
3-Fluorobenzoic acid 98%
3-Fluorobenzoic acid
Formula: C₇H₅FO₂
Melting Pt: 121…125 °C
Density: 1.474 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002489
CAS Number: 455-38-9
EINECS: 207-248-0