4-Fluorobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC3520-100G PC3520-1KG PC3520-500G
APOSPC3520-100GEA 21.9 EUR
APOSPC3520-100G APOSPC3520-1KG APOSPC3520-500G
4-Fluorobenzoic acid 98%
4-Fluorobenzoic acid
Formula: C₇H₅FO₂
Melting Pt: 182…187 °C
Density: 1.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002530
CAS Number: 456-22-4
EINECS: 207-259-0