2-Bromo-3'-chloro-4'-fluoroacetophenone 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Chloro-4-fluorophenacyl bromide
PC3542-1G PC3542-25G PC3542-5G
APOSPC3542-1GEA 46.4 EUR
APOSPC3542-1G APOSPC3542-25G APOSPC3542-5G
2-Bromo-3'-chloro-4'-fluoroacetophenone 98%
2-Bromo-3'-chloro-4'-fluoroacetophenone
Formula: C₈H₅BrClFO
MW: 251.48 g/mol
Melting Pt: 35…37 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05154991
CAS Number: 63529-30-6
UN: 2811
ADR: 6.1 III