2-Fluoro-5-iodobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC3543-5G PC3543-25G PC3543-100G
APOSPC3543-5GEA 23.2 EUR
APOSPC3543-5G APOSPC3543-25G APOSPC3543-100G
2-Fluoro-5-iodobenzonitrile 98%
2-Fluoro-5-iodobenzonitrile
Formula: C₇H₃FIN
MW: 247.01 g/mol
Melting Pt: 71…73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094225
CAS Number: 351003-36-6
UN: 3439
ADR: 6.1,III