2-Fluoro-5-nitroaniline 96%
Supplier: Apollo Scientific
Warning
PC3549-5G PC3549-1G
APOSPC3549-5GEA 0 EUR
APOSPC3549-5G APOSPC3549-1G
2-Fluoro-5-nitroaniline 96%
2-Fluoro-5-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 97…103 °C
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007652
CAS Number: 369-36-8
EINECS: 206-720-3
UN: 2811
ADR: 6.1 III