4-Fluorobenzonitrile 99%
Supplier: Apollo Scientific
Danger
Synonyms: p-Fluorobenzonitrile
PC3550-100G PC3550-500G
APOSPC3550-100GEA 24.8 EUR
APOSPC3550-100G APOSPC3550-500G
4-Fluorobenzonitrile 99%
4-Fluorobenzonitrile
Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 188 °C/750mmHg
Melting Pt: 35…37 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00001812
CAS Number: 1194-02-1
EINECS: 214-784-9
UN: 1325
ADR: 4.1 II