4-Fluoro-3-(trifluoromethyl)benzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 4-Fluoro-3-(trifluoromethyl)benzene-1-sulphonyl chloride
PC3552-25G PC3552-5G PC3552-100G
APOSPC3552-25GEA 55.1 EUR
APOSPC3552-25G APOSPC3552-5G APOSPC3552-100G
4-Fluoro-3-(trifluoromethyl)benzenesulphonyl chloride
4-Fluoro-3-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃ClF₄O₂S
MW: 262.61 g/mol
Boiling Pt: 92 °C/0.8mmHg
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01091001
CAS Number: 1682-10-6
UN: 3265
ADR: 8 III