1-Phenyl-5-(trifluoromethyl)pyrazole-4-carbonylchloride 97%
Supplier: Apollo Scientific
Danger
PC3554-10G PC3554-1G PC3554-5G
APOSPC3554-10GEA 288 EUR
APOSPC3554-10G APOSPC3554-1G APOSPC3554-5G
1-Phenyl-5-(trifluoromethyl)pyrazole-4-carbonylchloride 97%
1-Phenyl-5-(trifluoromethyl)pyrazole-4-carbonylchloride
Formula: C₁₁H₆ClF₃N₂O
MW: 274.63 g/mol
Boiling Pt: 110 °C/0.2mmHg
Melting Pt: 40…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00068139
CAS Number: 175137-14-1
UN: 3261
ADR: 8,III