4-Fluorobenzophenone 98%
Supplier: Apollo Scientific
Warning
PC3570-1KG PC3570-250G PC3570-50G
APOSPC3570-1KGEA 321 EUR
APOSPC3570-1KG APOSPC3570-250G APOSPC3570-50G
4-Fluorobenzophenone 98%
4-Fluorobenzophenone
Formula: C₁₃H₉FO
MW: 200.21 g/mol
Boiling Pt: 130…132 °C/3mmHg
Melting Pt: 45…49 °C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000352
CAS Number: 345-83-5
EINECS: 206-463-7