2-Fluoro-α,α,α-trichlorotoluene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-alpha,alpha,alpha-trichlorotoluene, 2-Fluorobenzotrichloride, o-Fluoro-α,α,α-trichlorotoluene
PC3578-25G PC3578-100G PC3578-5G
APOSPC3578-25GEA 108 EUR
APOSPC3578-25G APOSPC3578-100G APOSPC3578-5G
2-Fluoro-α,α,α-trichlorotoluene 99%
2-Fluoro-α,α,α-trichlorotoluene
Formula: C₇H₄Cl₃F
MW: 213.47 g/mol
Boiling Pt: 75 °C/5mmHg
Melting Pt: –20 °C
Density: 1.453 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000787
CAS Number: 488-98-2
EINECS: 207-692-5