Methyl-4-bromo-3-fluorobenzoate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 4-bromo-3-fluorobenzoate
PC3584-100G
PC3584-25G
PC3584-5G
APOSPC3584-100GEA
335
EUR
APOSPC3584-100G
APOSPC3584-25G
APOSPC3584-5G
Methyl-4-bromo-3-fluorobenzoate 98%
Methyl 4-bromo-3-fluorobenzoate
Formula:
C₈H₆BrFO₂ MW: 233.04 g/mol Melting Pt: 64…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD08436044 CAS Number: 849758-12-9 |