α,α,α,3-Tetrafluorotoluene 99%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Fluorobenzotrifluoride, 1-Fluoro-3-(trifluoromethyl)benzene
PC3590-100G PC3590-250G PC3590-1KG PC3590-25G
APOSPC3590-100GEA 45.1 EUR
APOSPC3590-100G APOSPC3590-250G APOSPC3590-1KG APOSPC3590-25G
α,α,α,3-Tetrafluorotoluene 99%
α,α,α,3-Tetrafluorotoluene
Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 101…102 °C
Density: 1.302 g/cm³ (25 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00000382
CAS Number: 401-80-9
EINECS: 206-933-1
UN: 1993
ADR: 3,II