α,α,α,4-Tetrafluorotoluene 99%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Fluorobenzotrifluoride, 1-Fluoro-4-(trifluoromethyl)benzene
PC3600-100G PC3600-25G
APOSPC3600-100GEA 43.5 EUR
APOSPC3600-100G APOSPC3600-25G
α,α,α,4-Tetrafluorotoluene 99%
α,α,α,4-Tetrafluorotoluene
Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 102…105 °C
Melting Pt: –42…–41 °C
Density: 1.293 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000399
CAS Number: 402-44-8
EINECS: 206-944-1
UN: 1993
ADR: 3,II