3-Methyl-5-(trifluoromethoxy)benzyl bromide 98%
Supplier: Apollo Scientific
Danger
PC3604-1G PC3604-5G
APOSPC3604-1GEA 69.6 EUR
APOSPC3604-1G APOSPC3604-5G
3-Methyl-5-(trifluoromethoxy)benzyl bromide 98%
3-Methyl-5-(trifluoromethoxy)benzyl bromide
Formula: C₉H₈BrF₃O
MW: 269.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08741403
CAS Number: 916420-55-8
UN: 3265
ADR: 8 II