3-Fluorobenzoyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Fluorobenzoylchloride, m-Fluorobenzoyl chloride
PC3620-100G PC3620-25G
APOSPC3620-100GEA 48 EUR
APOSPC3620-100G APOSPC3620-25G
3-Fluorobenzoyl chloride 98%
3-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
Boiling Pt: 189 °C
Melting Pt: –30 °C
Density: 1.32 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000670
CAS Number: 1711-07-5
EINECS: 216-977-3
UN: 3265
ADR: 8,II