4-Fluorobenzoyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: p-Fluorobenzoyl chloride
PC3630-100G PC3630-1KG PC3630-500G
APOSPC3630-100GEA 26.1 EUR
APOSPC3630-100G APOSPC3630-1KG APOSPC3630-500G
4-Fluorobenzoyl chloride 98%
4-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 82 °C/20mmHg
Melting Pt: 44540 °C
Density: 1.326 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000684
CAS Number: 403-43-0
EINECS: 206-961-4
UN: 3265
ADR: 8,II