1-(3-Chloro-4-fluorophenyl)pyrrolidine
Supplier: Apollo Scientific
PC3653-1G PC3653-5G
APOSPC3653-1GEA 132 EUR
APOSPC3653-1G APOSPC3653-5G
1-(3-Chloro-4-fluorophenyl)pyrrolidine
1-(3-Chloro-4-fluorophenyl)pyrrolidine
Formula: C₁₀H₁₁ClFN
Storage Temperature: Ambient
MDL Number: MFCD09878399
CAS Number: 1000339-33-2