2,5-Difluoropyridine 96%
Supplier: Apollo Scientific
Warning
PC3685-25G PC3685-5G
APOSPC3685-25GEA 125 EUR
APOSPC3685-25G APOSPC3685-5G
2,5-Difluoropyridine 96%
2,5-Difluoropyridine
Formula: C₅H₃F₂N
Boiling Pt: 115 °C
Melting Pt: –33 °C
Density: 1.274 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD04114191
CAS Number: 84476-99-3
UN: 1993
ADR: 3,III