α-Bromo-3-fluorotoulene 96%
Supplier: Apollo Scientific
Danger
Synonyms: α-Bromo-3-fluorotoluene, α-Bromo-m-fluorotoulene, alpha-Bromo-m-fluorotoulene, 1-(Bromomethyl)-3-fluorobenzene, m-Fluorobenzyl bromide, 3-Fluorobenzyl bromide
PC3694-100G PC3694-250G PC3694-25G
APOSPC3694-100GEA 43.5 EUR
APOSPC3694-100G APOSPC3694-250G APOSPC3694-25G
α-Bromo-3-fluorotoulene 96%
α-Bromo-3-fluorotoulene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 88 °C/20mmHg
Density: 1.541 g/cm³ (25 °C)
Flash Pt: 61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000340
CAS Number: 456-41-7
EINECS: 207-263-2
UN: 3265
ADR: 8 II