2,4,6-Trifluorobenzenesulphonamide
Supplier: Apollo Scientific
Warning
PC3697-1G PC3697-5G
APOSPC3697-1GEA 63.8 EUR
APOSPC3697-1G APOSPC3697-5G
2,4,6-Trifluorobenzenesulphonamide
2,4,6-Trifluorobenzenesulphonamide
Formula: C₆H₄F₃NO₂S
Melting Pt: 109…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01940428
CAS Number: 502642-77-5