5-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester 97%
Supplier: Apollo Scientific
Synonyms: Methyl 5-(trifluoromethyl)picolinate, Methyl 5-(trifluoromethyl)pyridine-2-carboxylate
PC3708-10G PC3708-25G
APOSPC3708-10GEA 190 EUR
APOSPC3708-10G APOSPC3708-25G
5-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester 97%
5-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester
Formula: C₈H₆F₃NO₂
MW: 205.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08741523
CAS Number: 124236-37-9