4-Fluorobenzyl chloride 99%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Fluorobenzyl chloride, p-Fluorobenzyl chloride, 1-Chloromethyl-4-fluorobenzene, α-Chloro-4-fluorotoluene, 1-(Chloromethyl)-4-fluorobenzene
PC3720-100G PC3720-500G
APOSPC3720-100GEA 21.9 EUR
APOSPC3720-100G APOSPC3720-500G
4-Fluorobenzyl chloride 99%
4-Fluorobenzyl chloride
Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 185 °C
Melting Pt: –18 °C
Density: 1.207 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00000913
CAS Number: 352-11-4
EINECS: 206-516-4
UN: 2920
ADR: 8+3 II