2-Fluorobiphenyl 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-1,1'-biphenyl, PFB No. 1
PC3724-25G PC3724-5G PC3724-100G
APOSPC3724-25GEA 24.8 EUR
APOSPC3724-25G APOSPC3724-5G APOSPC3724-100G
2-Fluorobiphenyl 98%
2-Fluorobiphenyl
Formula: C₁₂H₉F
MW: 172.2 g/mol
Boiling Pt: 248 °C
Melting Pt: 71…76 °C
Density: 1.245 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000317
CAS Number: 321-60-8
EINECS: 206-290-7
UN: 3077
ADR: 9+EHS,III