4-Fluorobiphenyl 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
PFB No. 3, 4-Fluoro-1,1-biphenyl
PC3726-5G
PC3726-25G
APOSPC3726-5GEA
72.5
EUR
APOSPC3726-5G
APOSPC3726-25G
4-Fluorobiphenyl 97%
4-Fluorobiphenyl
Formula:
C₁₂H₉F MW: 172.2 g/mol Boiling Pt: 310 °C Melting Pt: 74…79 °C Density: 1.288 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011650 CAS Number: 324-74-3 EINECS: 206-304-1 |