2-Fluorocinnamic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(2-Fluorophenyl)propenoic acid
PC3730-25G PC3730-5G
APOSPC3730-25GEA 147 EUR
APOSPC3730-25G APOSPC3730-5G
2-Fluorocinnamic acid 98%
2-Fluorocinnamic acid
Formula: C₉H₇FO₂
MW: 166.15 g/mol
Melting Pt: 178…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00004370
CAS Number: 451-69-4
EINECS: 207-195-3