5-Fluorodihydro-6-hydroxy-2,4(1H,3H)pyrimidinedione monohydrate 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Fluoro-6-hydroxyhydrouracil monohydrate
PC3745-5G PC3745-1G PC3745-25G
APOSPC3745-5GEA 116 EUR
APOSPC3745-5G APOSPC3745-1G APOSPC3745-25G
5-Fluorodihydro-6-hydroxy-2,4(1H,3H)pyrimidinedione monohydrate 98%
5-Fluorodihydro-6-hydroxy-2,4(1H,3H)pyrimidinedione monohydrate
Formula: C₄H₇FN₂O₄
MW: 166.11 g/mol
Melting Pt: >130 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00150905
CAS Number: 37103-91-6
UN: 2811
ADR: 6.1,III