4'-Fluoro-2'-hydroxyacetophenone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Fluoro-2-hydroxyacetophenone
PC3755P-100G
PC3755P-25G
APOSPC3755P-100GEA
296
EUR
APOSPC3755P-100G
APOSPC3755P-25G
4'-Fluoro-2'-hydroxyacetophenone 98%
4'-Fluoro-2'-hydroxyacetophenone
Formula:
C₈H₇FO₂ MW: 154.14 g/mol Boiling Pt: 65 °C/2mmHg Melting Pt: 32…35 °C Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143203 CAS Number: 1481-27-2 |