5-Fluoro-1-indanone
Supplier: Apollo Scientific
Warning
Synonyms: 5-Fluoro-2,3-dihydro-1H-inden-1-one
PC3757-100G PC3757-25G
APOSPC3757-100GEA 266 EUR
APOSPC3757-100G APOSPC3757-25G
5-Fluoro-1-indanone
5-Fluoro-1-indanone
Formula: C₉H₇FO
MW: 150.15 g/mol
Boiling Pt: 92…93 °C/3mmHg
Melting Pt: 37…40 °C
Density: 1.216 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00041031
CAS Number: 700-84-5