5-Fluoroindole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Fluoro-1H-indole
PC3758-5G PC3758-1G
APOSPC3758-5GEA 30.6 EUR
APOSPC3758-5G APOSPC3758-1G
5-Fluoroindole 98%
5-Fluoroindole
Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 45…48 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005671
CAS Number: 399-52-0
EINECS: 206-917-4