6-(Trifluoromethyl)-4-quinolinol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydroxy-6-(trifluoromethyl)quinoline
PC3759-1G PC3759-5G
APOSPC3759-1GEA 21.9 EUR
APOSPC3759-1G APOSPC3759-5G
6-(Trifluoromethyl)-4-quinolinol 98%
6-(Trifluoromethyl)-4-quinolinol
Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
Melting Pt: 265…268 °C
Storage Temperature: Ambient
MDL Number: MFCD00209673
CAS Number: 49713-51-1