1-Fluoro-2-iodobenzene 99%
Supplier: Apollo Scientific
Danger
PC3760-100G PC3760-500G PC3760-25G
APOSPC3760-100GEA 43.5 EUR
APOSPC3760-100G APOSPC3760-500G APOSPC3760-25G
1-Fluoro-2-iodobenzene 99%
1-Fluoro-2-iodobenzene
Formula: C₆H₄FI
Boiling Pt: 188…189 °C
Melting Pt: –41 to –40 °C
Density: 1.903 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00001031
CAS Number: 348-52-7
EINECS: 206-477-3
UN: 3082
ADR: 9+EHS,III