4,4,5,5,6,6,6-Heptafluorohexane-1,2-diol 97%
Supplier: Apollo Scientific
Warning
PC3763-1G PC3763-5G
APOSPC3763-1GEA 105 EUR
APOSPC3763-1G APOSPC3763-5G
4,4,5,5,6,6,6-Heptafluorohexane-1,2-diol 97%
4,4,5,5,6,6,6-Heptafluorohexane-1,2-diol
Formula: C₆H₇F₇O₂
Boiling Pt: 114 °C/22mmHg
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03424477
CAS Number: 1992-91-2