1-Fluoro-3-iodobenzene 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Fluoroiodobenzene
PC3770-100G
PC3770-25G
APOSPC3770-100GEA
88.6
EUR
APOSPC3770-100G
APOSPC3770-25G
1-Fluoro-3-iodobenzene 99%
1-Fluoro-3-iodobenzene
Formula:
C₆H₄FI Boiling Pt: 77…78 °C/19mmHg Density: 1.89 g/cm³ (25 °C) Flash Pt: 67 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001044 CAS Number: 1121-86-4 |