Difluoroacetic anhydride 97%
Supplier: Apollo Scientific
Danger
PC3791-25G PC3791-5G PC3791-100G
APOSPC3791-25GEA 58 EUR
APOSPC3791-25G APOSPC3791-5G APOSPC3791-100G
Difluoroacetic anhydride 97%
Difluoroacetic anhydride
Formula: C₄H₂F₄O₃
Boiling Pt: 125…127 °C
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02093315
CAS Number: 401-67-2
UN: 3265
ADR: 8 III