3-Fluoro-4-methoxybenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC3798-25G PC3798-5G PC3798-100G
APOSPC3798-25GEA 52.2 EUR
APOSPC3798-25G APOSPC3798-5G APOSPC3798-100G
3-Fluoro-4-methoxybenzonitrile 98%
3-Fluoro-4-methoxybenzonitrile
Formula: C₈H₆FNO
MW: 151.14 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00215835
CAS Number: 331-62-4
UN: 3439
ADR: 6.1,III