3-Fluoro-4-methoxybenzyl bromide 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4-(Bromomethyl)-2-fluoro-1-methoxybenzene
PC3801-25G PC3801-5G PC3801-1G
APOSPC3801-25GEA 0 EUR
APOSPC3801-25G APOSPC3801-5G APOSPC3801-1G
3-Fluoro-4-methoxybenzyl bromide 97%
3-Fluoro-4-methoxybenzyl bromide
Formula: C₈H₈BrFO
MW: 219.05 g/mol
Melting Pt: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00671768
CAS Number: 331-61-3
UN: 3261
ADR: 8 II