3-Chloro-5-fluorobenzoic acid 97%
Supplier: Apollo Scientific
Warning
PC3809-100G PC3809-25G PC3809-5G
APOSPC3809-100GEA 458 EUR
APOSPC3809-100G APOSPC3809-25G APOSPC3809-5G
3-Chloro-5-fluorobenzoic acid 97%
3-Chloro-5-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 153…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00973904
CAS Number: 25026-64-6