4-Fluoro-2-iodotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-3-iodo-4-methylbenzene, 4-Fluoro-2-iodo-1-methylbenzene
PC3810-100G PC3810-25G
APOSPC3810-100GEA 115 EUR
APOSPC3810-100G APOSPC3810-25G
4-Fluoro-2-iodotoluene 98%
4-Fluoro-2-iodotoluene
Formula: C₇H₆FI
MW: 236.03 g/mol
Boiling Pt: 204…208 °C
Density: 1.81 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00013709
CAS Number: 13194-67-7
EINECS: 236-153-7