(±)-4-Fluoromandelic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluoromandelic Acid, (±)-4-Fluorophenylglycolic acid
PC3817A-25G PC3817A-1G PC3817A-5G
APOSPC3817A-25GEA 56.7 EUR
APOSPC3817A-25G APOSPC3817A-1G APOSPC3817A-5G
(±)-4-Fluoromandelic acid 98%
(±)-4-Fluoromandelic acid
Formula: C₈H₇FO₃
Melting Pt: 133…140 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00044402
CAS Number: 395-33-5
EINECS: 206-899-8