3'-Fluoro-4'-methoxyacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-Fluoro-4-methoxyphenyl)-1-ethanone
PC3817E-100G PC3817E-25G PC3817E-5G
APOSPC3817E-100GEA 119 EUR
APOSPC3817E-100G APOSPC3817E-25G APOSPC3817E-5G
3'-Fluoro-4'-methoxyacetophenone 98%
3'-Fluoro-4'-methoxyacetophenone
Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 147…148 °C/20mmHg
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00026219
CAS Number: 455-91-4
EINECS: 207-253-8