2-Fluoro-p-anisaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Fluoro-4-methoxybenzaldehyde, 2-Fluoro-4-anisaldehyde
PC3817EC-25G
PC3817EC-5G
APOSPC3817EC-25GEA
78.3
EUR
APOSPC3817EC-25G
APOSPC3817EC-5G
2-Fluoro-p-anisaldehyde 95%
2-Fluoro-p-anisaldehyde
Formula:
C₈H₇FO₂ MW: 154.14 g/mol Melting Pt: 43…48 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00236679 CAS Number: 331-64-6 |