6-Fluoro-m-anisaldehyde 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-5-methoxybenzaldehyde, 6-Fluoro-3-anisaldehyde
PC3817ED-100G PC3817ED-25G PC3817ED-5G
APOSPC3817ED-100GEA 398 EUR
APOSPC3817ED-100G APOSPC3817ED-25G APOSPC3817ED-5G
6-Fluoro-m-anisaldehyde 98%
6-Fluoro-m-anisaldehyde
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 80 °C/0.9mmHg
Density: 1.237 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00070795
CAS Number: 105728-90-3