2-Fluoro-6-methoxybenzonitrile 95%
Supplier: Apollo Scientific
|
Warning
|
PC3817F-100G
PC3817F-25G
APOSPC3817F-100GEA
109
EUR
APOSPC3817F-100G
APOSPC3817F-25G
2-Fluoro-6-methoxybenzonitrile 95%
2-Fluoro-6-methoxybenzonitrile
Formula:
C₈H₆FNO MW: 151.14 g/mol Boiling Pt: 245 Melting Pt: 102…104 °C Flash Pt: 94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042291 CAS Number: 94088-46-7 EINECS: 302-047-5 UN: 3439 ADR: 6.1 III |